Merck
  • Biochemical characterization and comparison of aspartylglucosaminidases secreted in venom of the parasitoid wasps Asobara tabida and Leptopilina heterotoma.
Biochemical characterization and comparison of aspartylglucosaminidases secreted in venom of the parasitoid wasps Asobara tabida and Leptopilina heterotoma.
PloS one (2017-07-26)
Quentin Coulette, Séverine Lemauf, Dominique Colinet, Geneviève Prévost, Caroline Anselme, Marylène Poirié, Jean-Luc Gatti
ZUSAMMENFASSUNG
Aspartylglucosaminidase (AGA) is a low-abundance intracellular enzyme that plays a key role in the last stage of glycoproteins degradation, and whose deficiency leads to human aspartylglucosaminuria, a lysosomal storage disease. Surprisingly, high amounts of AGA-like proteins are secreted in the venom of two phylogenetically distant hymenopteran parasitoid wasp species, Asobara tabida (Braconidae) and Leptopilina heterotoma (Cynipidae). These venom AGAs have a similar domain organization as mammalian AGAs. They share with them key residues for autocatalysis and activity, and the mature α- and β-subunits also form an (αβ)2 structure in solution. Interestingly, only one of these AGAs subunits (α for AtAGA and β for LhAGA) is glycosylated instead of the two subunits for lysosomal human AGA (hAGA), and these glycosylations are partially resistant to PGNase F treatment. The two venom AGAs are secreted as fully activated enzymes, they have a similar aspartylglucosaminidase activity and are both also efficient asparaginases. Once AGAs are injected into the larvae of the Drosophila melanogaster host, the asparaginase activity may play a role in modulating their physiology. Altogether, our data provide new elements for a better understanding of the secretion and the role of venom AGAs as virulence factors in the parasitoid wasps' success.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Kaninchen-IgG-(Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Glutamat-Oxalessig-Transaminase Zubereitung aus Schweineherz, Type I, ammonium sulfate suspension, 200-500 units/mg protein