Merck
Salmonella Typhimurium disrupts Sirt1/AMPK checkpoint control of mTOR to impair autophagy.
PLoS pathogens (2017-02-14)
Raja Ganesan, Nina Judith Hos, Saray Gutierrez, Julia Fischer, Joanna Magdalena Stepek, Evmorphia Daglidu, Martin Krönke, Nirmal Robinson
ZUSAMMENFASSUNG
During intracellular infections, autophagy significantly contributes to the elimination of pathogens, regulation of pro-inflammatory signaling, secretion of immune mediators and in coordinating the adaptive immune system. Intracellular pathogens such as S. Typhimurium have evolved mechanisms to circumvent autophagy. However, the regulatory mechanisms targeted by S. Typhimurium to modulate autophagy have not been fully resolved. Here we report that cytosolic energy loss during S. Typhimurium infection triggers transient activation of AMPK, an important checkpoint of mTOR activity and autophagy. The activation of AMPK is regulated by LKB1 in a cytosolic complex containing Sirt1 and LKB1, where Sirt1 is required for deacetylation and subsequent activation of LKB1. S. Typhimurium infection targets Sirt1, LKB1 and AMPK to lysosomes for rapid degradation resulting in the disruption of the AMPK-mediated regulation of mTOR and autophagy. The degradation of cytosolic Sirt1/LKB1/AMPK complex was not observed with two mutant strains of S. Typhimurium, ΔssrB and ΔssaV, both compromising the pathogenicity island 2 (SPI2). The results highlight virulence factor-dependent degradation of host cell proteins as a previously unrecognized strategy of S. Typhimurium to evade autophagy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bafilomycin A1 aus Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Pepstatin A, microbial, ≥90% (HPLC)
Sigma-Aldrich
Anti-LC3B in Kaninchen hergestellte Antikörper, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
Leptomycin B -Lösung aus Streptomyces sp., ≥95% (HPLC), Supplied in methanol: water (7:3)
Sigma-Aldrich
Lactacystin, ≥90% (HPLC)
Sigma-Aldrich
Anti-Matrix Metalloproteinase-21, Hinge Region antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution