Merck
Membrane Targeting of Disheveled Can Bypass the Need for Arrow/LRP5.
Scientific reports (2017-08-02)
Prameet Kaur, Vanessa Yuk Man Lam, Anirudh Gautam Mannava, Jahnavi Suresh, Andreas Jenny, Nicholas S Tolwinski
ZUSAMMENFASSUNG
The highly conserved Wnt signaling pathway regulates cell proliferation and differentiation in vertebrates and invertebrates. Upon binding of a Wnt ligand to a receptor of the Fz family, Disheveled (Dsh/Dvl) transduces the signal during canonical and non-canonical Wnt signaling. The specific details of how this process occurs have proven difficult to study, especially as Dsh appears to function as a switch between different branches of Wnt signaling. Here we focus on the membrane-proximal events that occur once Dsh is recruited to the membrane. We show that membrane-tethering of the Dsh protein is sufficient to induce canonical Wnt signaling activation even in the absence of the Wnt co-receptor Arrow/LRP5/6. We map the protein domains required for pathway activation in membrane tethered constructs finding that both the DEP and PDZ domains are dispensable for canonical signaling only in membrane-tethered Dsh, but not in untethered/normal Dsh. These data lead to a signal activation model, where Arrow is required to localize Dsh to the membrane during canonical Wnt signaling placing Dsh downstream of Arrow.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® BioM2 in Maus hergestellte Antikörper, clone M2, purified immunoglobulin, buffered aqueous glycerol solution