Merck
Endorepellin-evoked Autophagy Contributes to Angiostasis.
The Journal of biological chemistry (2016-07-21)
Atul Goyal, Maria A Gubbiotti, Daphney R Chery, Lin Han, Renato V Iozzo
ZUSAMMENFASSUNG
Endorepellin, the C-terminal domain of perlecan, is an angiostatic molecule that acts as a potent inducer of autophagy via its interaction with VEGFR2. In this study, we examined the effect of endorepellin on endothelial cells using atomic force microscopy. Soluble endorepellin caused morphological and biophysical changes such as an increase in cell surface roughness and cell height. Surprisingly, these changes were not accompanied by alterations in the endothelial cell elastic modulus. We discovered that endorepellin-induced autophagic flux led to co-localization of mammalian target of rapamycin with LC3-positive autophagosomes. Endorepellin functioned upstream of AMP-activated kinase α, as compound C, an inhibitor of AMP-activated kinase α, abrogated endorepellin-mediated activation and co-localization of Beclin 1 and LC3, thereby reducing autophagic progression. Functionally, we discovered that both endorepellin and Torin 1, a canonical autophagic inducer, blunted ex vivo angiogenesis. We conclude that autophagy is a novel mechanism by which endorepellin promotes angiostasis independent of nutrient deprivation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-LC3B in Kaninchen hergestellte Antikörper, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Kaninchen-IgG-Antikörper der Ziege, HRP-konjugiert, 1 mg/mL, Upstate®