Merck
  • Dimerization of the transmembrane domain of amyloid precursor protein is determined by residues around the γ-secretase cleavage sites.
Dimerization of the transmembrane domain of amyloid precursor protein is determined by residues around the γ-secretase cleavage sites.
The Journal of biological chemistry (2017-08-10)
Yan Yan, Ting-Hai Xu, Kaleeckal G Harikumar, Laurence J Miller, Karsten Melcher, H Eric Xu
ZUSAMMENFASSUNG
One of the hallmarks of Alzheimer's disease is the formation of extracellular amyloid plaques that consist mainly of abnormally aggregated forms of amyloid β (Aβ) peptides. These peptides are generated by γ-secretase-catalyzed cleavage of a dimeric membrane-bound C-terminal fragment (C99) of the amyloid precursor protein. Although C99 homodimerization has been linked to Aβ production and changes in the aggregation-determining Aβ42/Aβ40 ratio, the motif through which C99 dimerizes has remained controversial. Here, we have used two independent assays to gain insight into C99 homodimerization in the context of the membrane of live cells: bioluminescence resonance energy transfer and Tango membrane protein-protein interaction assays, which were further confirmed by traditional pull-down assays. Our results indicate a four-amino acid region within the C99 transmembrane helix (
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonale ANTI-FLAG® M2-Peroxidase (HRP) in Maus hergestellte Antikörper, clone M2, purified immunoglobulin, buffered aqueous glycerol solution