Merck
A Living Biobank of Breast Cancer Organoids Captures Disease Heterogeneity.
Cell (2017-12-12)
Norman Sachs, Joep de Ligt, Oded Kopper, Ewa Gogola, Gergana Bounova, Fleur Weeber, Anjali Vanita Balgobind, Karin Wind, Ana Gracanin, Harry Begthel, Jeroen Korving, Ruben van Boxtel, Alexandra Alves Duarte, Daphne Lelieveld, Arne van Hoeck, Robert Frans Ernst, Francis Blokzijl, Isaac Johannes Nijman, Marlous Hoogstraat, Marieke van de Ven, David Anthony Egan, Vittoria Zinzalla, Jurgen Moll, Sylvia Fernandez Boj, Emile Eugene Voest, Lodewyk Wessels, Paul Joannes van Diest, Sven Rottenberg, Robert Gerhardus Jacob Vries, Edwin Cuppen, Hans Clevers
ZUSAMMENFASSUNG
Breast cancer (BC) comprises multiple distinct subtypes that differ genetically, pathologically, and clinically. Here, we describe a robust protocol for long-term culturing of human mammary epithelial organoids. Using this protocol, >100 primary and metastatic BC organoid lines were generated, broadly recapitulating the diversity of the disease. BC organoid morphologies typically matched the histopathology, hormone receptor status, and HER2 status of the original tumor. DNA copy number variations as well as sequence changes were consistent within tumor-organoid pairs and largely retained even after extended passaging. BC organoids furthermore populated all major gene-expression-based classification groups and allowed in vitro drug screens that were consistent with in vivo xeno-transplantations and patient response. This study describes a representative collection of well-characterized BC organoids available for cancer research and drug development, as well as a strategy to assess in vitro drug response in a personalized fashion.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tamoxifen, ≥99%
Roche
Lysepuffer für Erythrozyten, solution, Roche, pkg of 100 mL, sufficient for 50-500 reactions
Sigma-Aldrich
SB 202190, ≥98% (HPLC)
Sigma-Aldrich
Collagenase aus Clostridium histolyticum, powder, Suitable for the digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
Sigma-Aldrich
Nutlin-3, ≥98% (HPLC), powder