Merck
  • Simultaneous determination of volatile organic compounds with a wide range of polarities in urine by headspace solid-phase microextraction coupled to gas chromatography/mass spectrometry.
Simultaneous determination of volatile organic compounds with a wide range of polarities in urine by headspace solid-phase microextraction coupled to gas chromatography/mass spectrometry.
Rapid communications in mass spectrometry : RCM (2017-01-14)
Han-Na Song, Chong Hyeak Kim, Won-Yong Lee, Sung-Hee Cho
ZUSAMMENFASSUNG
Volatile organic compounds (VOCs) are ubiquitous environmental pollutants that have a high vapor pressure at room temperature. Some VOCs have been classified as carcinogenic to humans by the International Agency for Research on Cancer (IARC), because they can bind to DNA and cause cell mutations. Therefore, monitoring of VOCs in human urine is very important to evaluate the correlation between exposure to VOCs and human disease. We have developed an improved analytical method for the simultaneous determination of VOCs with a wide range of polarities in human urine samples by headspace solid-phase microextraction (HS-SPME) coupled to gas chromatography/mass spectrometry (GC/MS). In the improved method, a bi-polar carboxen-polydimethylsiloxane (CAR/PDMS) fiber was used for the optimized extraction of 15 VOCs with a wide range of polarities, including benzene, toluene, ethylbenzene, xylenes (BTEX), alkylbenzenes, cresols, and naphthalene, in human urine samples. Extracted VOCs from the human urine were effectively separated by GC using a mid-polarity column (DB-35, 35% phenylmethylpolysiloxane) and monitored by MS using extracted ion monitoring (EIM) mode. Under the optimized method, the linearity of the calibration curves was greater than 0.993. The limits of detection (LODs) at a signal-to-noise (S/N) ratio of 3 were 0.3-0.6 ng/mL. The coefficients of variation were in the range of 0.1-9.7% for within-day variation and 0.2-14.2% for day-to-day variation. The method was shown to be rapid and simple for the simultaneous determination of VOCs with a wide range of polarities in human urine and it could be applied to monitoring and to biomedical investigations to check exposure to VOCs. Copyright © 2017 John Wiley & Sons, Ltd.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Supelco
ORBO Carboxen-564 Röhrchen, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 78 HBr auf Carboxen® 564 speziell gereinigt (20/45), 400/200 mg, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
Supelco
Carboxen®-1010 PLOT Kapillar-GC-Säule, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Supelco
Carboxen® Adsorptionsmittel, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, black hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
Carboxen®-1006 PLOT Kapillar-GC-Säule, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, black hub
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
Carboxen®-1010 PLOT Kapillar-GC-Säule, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, light blue hub
Supelco
SPME-Faseranordnung Polyacrylat (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
Tragbarer SPME-Field-Sampler, coating PDMS/DVB
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
SPME-Faseranordnung Polyacrylat (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, blue hub
Supelco
Tragbarer SPME-Field-Sampler, coating PDMS
Supelco
ORBO-93 Sorbensröhrchen, ORBO tube I.D. × L 6 mm × 95 mm, Bed A 180 mg, Bed B 90 mg, 60/80 mesh, pkg of 25 ea