Merck
  • Electrodeposition of self-assembled poly(3,4-ethylenedioxythiophene) @gold nanoparticles on stainless steel wires for the headspace solid-phase microextraction and gas chromatographic determination of several polycyclic aromatic hydrocarbons.
Electrodeposition of self-assembled poly(3,4-ethylenedioxythiophene) @gold nanoparticles on stainless steel wires for the headspace solid-phase microextraction and gas chromatographic determination of several polycyclic aromatic hydrocarbons.
Journal of chromatography. A (2016-10-22)
Liu Yang, Jie Zhang, Faqiong Zhao, Baizhao Zeng
ZUSAMMENFASSUNG
In this work, a novel poly(3,4-ethylenedioxythiophene)@Au nanoparticles (PEDOT@AuNPs) hybrid coating was prepared and characterized. Firstly, the monomer 3,4-ethylenedioxythiophene was self-assembled on AuNPs, and then electropolymerization was performed on a stainless steel wire by cyclic voltammetry. The obtained PEDOT@AuNPs coating was rough and showed cauliflower-like micro-structure with thickness of ∼40μm. It displayed high thermal stability (up to 330°C) and mechanical stability and could be used for at least 160 times of solid phase microextraction (SPME) without decrease of extraction performance. The coating exhibited high extraction capacity for some environmental pollutants (e.g. naphthalene, 2-methylnaphthalene, acenaphthene, fluorene and phenathrene) due to the hydrophobic interaction between the analytes and PEDOT and the additional physicochemical affinity between polycyclic aromatic hydrocarbons and AuNPs. Through coupling with GC detection, good linearity (correlation coefficients higher than 0.9894), wide linear range (0.01-100μgL
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, black hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, black hub
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, light blue hub
Supelco
SPME-Faseranordnung Polyacrylat (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
Tragbarer SPME-Field-Sampler, coating PDMS/DVB
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME-Faseranordnung Polyacrylat (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
Tragbarer SPME-Field-Sampler, coating PDMS
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, black hub
Supelco
SPME StableFlex Methodenentwicklungs-Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 4
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
SPME StableFlex Methodenentwicklungs-Kit, Assortment, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
Tragbarer SPME-Field-Sampler, coating CAR/PDMS
Supelco
SPME-Faseranordnung, Carbowax-Polyethylenglycol (PEG)-Beschichtung, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 3, purple hub
Supelco
SPME-Fasersortiment-Kit 4, Assortment #4, Fused Silica (1 cm), size 24 ga, Manual Holder, pk of 3