Merck
Heart failure, aging, and renal synthesis of dopamine.
American journal of kidney diseases : the official journal of the National Kidney Foundation (2001-09-05)
A Ferreira, P Bettencourt, M Pestana, F Correia, P Serrão, L Martins, M Cerqueira-Gomes, P Soares-Da-Silva
ABSTRACT
The present study evaluates renal dopaminergic activity in 23 patients with heart failure (HF), 10 age-matched controls, and 10 young subjects during normal-salt (NS) intake and after 8 days of low-salt (LS) intake (patients with HF and age-matched controls only). LS intake produced a marked reduction in urine volume in patients with HF but failed to affect urine volume in age-matched controls. Urinary sodium and fractional excretion of sodium were markedly reduced by LS intake in patients with HF and age-matched controls. Daily urinary excretion of L-3,4-dihydroxyphenylalanine (L-dopa) and dopamine was lower in patients with HF than in age-matched controls. LS intake failed to alter L-dopa and dopamine urinary excretion in control subjects. In patients with HF, LS intake produced a significant decrease in urinary L-dopa excretion, but failed to alter the urinary excretion of dopamine. No significant differences were observed in urinary L-dopa, dopamine, and dopamine metabolite levels between aged controls and young healthy subjects. Urinary dopamine-L-dopa ratios in patients with HF on LS intake (24.5 +/- 7.1) were significantly greater than those with NS intake (11.6 +/- 1.3). Urinary dopamine-L-dopa ratios in old control subjects (LS, 9.7 +/- 1.3; NS, 9.3 +/- 1.1) did not differ from those in young healthy subjects (9.2 +/- 0.8). LS intake produced a marked increase in plasma aldosterone levels in both patients with HF (84.6 +/- 14.4 to 148.2 +/- 20.4 pg/mL; P = 0.0008) and controls (102.1 +/- 13.4 to 151.6 +/- 15.7 pg/mL; P < 0.04). Plasma norepinephrine levels were not significantly affected by LS intake in controls (5.1 +/- 1.62 to 6.3 +/- 1.6 pmol/mL; P = 0.22), but were significantly increased in patients with HF (5.8 +/- 0.8 to 7.1 +/- 0.9 pmol/mL; P = 0.04). In conclusion, patients with HF are endowed with an enhanced ability to take up (or decarboxylate) filtered L-dopa, which might counterbalance the reduced renal delivery of L-dopa, contributing to a relative preservation of dopamine synthesis. This may result as a compensatory mechanism, activated by stimuli leading to sodium reabsorption. Age seems to have no influence on renal dopamine production.
Product Description
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
Taurine, ≥98%, FG
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
SAFC
L-Isoleucine
SAFC
Taurine
SAFC
L-Methionine
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%