Merck
  • The metabolic serine hydrolases and their functions in mammalian physiology and disease.
The metabolic serine hydrolases and their functions in mammalian physiology and disease.
Chemical reviews (2011-06-24)
Jonathan Z Long, Benjamin F Cravatt
Product Description
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland