Merck
  • TUDCA Promotes Phagocytosis by Retinal Pigment Epithelium via MerTK Activation.
TUDCA Promotes Phagocytosis by Retinal Pigment Epithelium via MerTK Activation.
Investigative ophthalmology & visual science (2015-03-26)
Hiromi Murase, Kazuhiro Tsuruma, Masamitsu Shimazawa, Hideaki Hara
ABSTRACT
Renewal and elimination of the aged photoreceptor outer segment (POS) by RPE cells is a daily rhythmic process that is important for long-term vision. Phagocytic dysfunction results in photoreceptor cell death. Tauroursodeoxycholic acid (TUDCA), an endogenous bile acid, is known to show neuroprotective effects in stroke, neurological diseases, and retinal degeneration models. In this study, we investigated the effects of TUDCA on retinal phagocytosis. We used pHrodo-succinimidyl ester (SE), a pH-sensitive fluorescent dye, to label the POS for monitoring phagocytosis. After ingestion, the intensity of pHrodo fluorescence increases because of the pH changes inside the liposome. An RPE cell line, ARPE-19, and primary human RPE cells were used to investigate the hydrogen peroxide (H2O2)-induced disruption of phagocytosis in the pH-sensitive fluorescence POS phagocytosis assay. Additionally, we examined whether TUDCA could promote phagocytic function. The intensity of pHrodo light emission increased in a time-dependent manner. Tauroursodeoxycholic acid enhanced phagocytosis of POS and protected against H2O2-induced phagocytic dysfunction. It also promoted phagocytic function via activation of Mer tyrosine kinase receptor (MerTK), which is known to have a key role in the physiological renewal of POS. These results suggest that TUDCA activates MerTK, which is important for phagocytosis of POS. Tauroursodeoxycholic acid may represent a new therapeutic option for the treatment of retinal diseases.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture