MilliporeSigma
Karl Fischer Reagents
Accurate water determination with Karl Fischer titration is an important analytical method used in many industrial segments like Pharma, Food & Beverages, Cosmetics, Petrochemicals and Technical industries to ensure high quality of the products they produce. Using our Aquastar® Karl Fischer reagents and standards the water content of solids, liquids and gas samples can be determined fast, easily and with a high degree of accuracy. Our sophisticated, high-quality Aquastar® portfolio comprises of volumetric and coulometric Karl Fischer reagents and appropriate water standards, Certified Reference Materials (CRMs) according to ISO 17034.
Products
Two amber glass bottles with white labels, featuring black text and green accents, capped with black lids, against a white background, typically used for pharmaceuticals or chemical reagents.. Aquastar® Reagents for Volumetric Karl Fischer Titration
Aquastar® Reagents for Volumetric Karl Fischer Titration
The volumetric Karl Fischer Titration method is used to determine the water content accurately by adding iodine-containing titrants to the sample. The sample needs to be dissolved in an appropriate solvent to release the water completely. This method can be used to determine water contents from 0.01 % up to 100 %, and promotes fast, precise, and reproducible results. With our Aquastar® Karl Fischer Reagents, we offer a complete range of high-quality volumetric Karl Fischer Reagents, one- and two-component reagents, and some specialties for specific samples. All titrants and solvents do not contain chloroform.
  • Aquastar® Volumetric Reagents – Advantages
    • High accuracy and excellent precision
    • Reagents for all types of applications
    • Rapid titration and reproducible results
    • No crystallization
    • Innovative formulation for special applications
    • Fast conditioning and stable drift
  • One-Component Reagents – the one-component titration method has all the reactive compounds such as iodine, base, sulfur dioxide, and the appropriate alcohol required for the Karl Fischer reaction in one reagent, the titrantAquastar® CombiTitrants are available in different concentrations to determine higher and lower water contents precisely. Aquastar® CombiMethanol and Aquastar® CombiSolvent are solvents typically used with one-component titrants to dissolve the sample. Our Aquastar® CombiMethanol is perfectly designed for Karl Fischer Titration, it has a low water content of 0.01% and is free of impurities to avoid side reactions. The Aquastar® CombiSolvent is a methanol-free solvent and is usable for all samples which dissolve in ethanol. The one-component titration can be used for all kinds of samples but is preferred for the titration of frequently changing sample matrices and for samples that require the addition of a solubilizer.
  • Two-Component Reagents – in a two-component system, the titrant is comprised of iodine and alcohol, whereas sulfur dioxide and a base are dissolved in the solvent. Aquastar® two component Titrants must be used along with Aquastar® two-component Solvent. Two different titrant concentrations are available for higher and lower water content. The two-component Karl Fischer volumetric method shows improved buffer capacity, faster titration rates, and slightly better titer stability. They are often used for series from the same sample matrices or for strong acidic or alkaline samples.
  • Reagents for Aldehyde and Ketones – Aquastar® CombiTitrant 5 Keto and Aquastar® CombiSolvent Keto reagents are necessary when performing Karl Fischer titration for water analysis in ketone and aldehyde samples. They cause side reactions when using standard reagents with methanol as solvent. Therefore, they are methanol-free to prevent side reactions and unreliable endpoints.
  • Special reagents for oils and fats – oils, fats and long-chain hydrocarbons are usually insoluble in methanol. Additional solubilizers are required to dissolve such samples for the complete release of water. Choose our Aquastar® CombiSolvent Oil, CombiSolvent Fats, or Solvent Oils & Fats reagents for such samples to determine their water contents accurately. They come with additional solubilizer to dissolve samples like mineral oils, kerosene, fuel, vegetable oils, fats, cremes or ointments for Karl Fischer Titration.
Two amber glass bottles with white labels, featuring black text and green accents, capped with black lids, against a white background, typically used for pharmaceuticals or chemical reagents. Aquastar® Reagents for Coulometric Karl Fischer Titration
Aquastar® Reagents for Coulometric Karl Fischer Titration
Coulometric Karl Fischer Titration is preferred for liquid samples with very low water content, typically in the range of 10 ppm to 10.000 ppm. It is a direct method, and the required iodine is generated during the titration process. Our distinguished Aquastar® range of Coulometric Karl Fischer reagents are available for titration cells with and without a diaphragm. All our coulometric reagents are chloroform-free.
  • Aquastar® Coulometric Reagents – Advantages
    • High accuracy and excellent precision
    • Rapid titration and reproducible results
    • Fast conditioning and stable drift
    • Only one reagent is necessary for anode and cathode chamber
  • Coulometric reagents for cells without a diaphragm – Our coulometric reagents Aquastar® CombiCoulomat fritless, and Aquastar® Anolyte can be used for coulometric Karl Fischer Titration for cells without a diaphragm.
  • Coulometric reagents for cells with diaphragm – the Aquastar® CombiCoulomat frit reagent is designed for cells with a diaphragm, while Aquastar®CombiCoulomat fritless reagent can be used in both cells with and without diaphragm. Both reagents contain all reaction components in a single solution and can be added in anode and cathode chamber. The benefit is, only one reagent is necessary.
  • Coulometric reagents for aldehyds and ketones – we also offer special methanol-free reagents for aldehyde and ketone samples. Our Aquastar® Anolyte K and Aquastar® Catholyte K reagents can be used to determine exact water content in aldehydes and ketones samples.
Four Aquastar water testing vials with green labels and measurement markings, designed for various stages of water quality analysis, displayed against a white background.
Aquastar® Water Standards
The Aquastar® portfolio contains a broad range of excellent water standards for titer determination, monitoring Karl Fischer equipment, and verifying measured results. Our Aquastar® Water Standards are CRMs (Certified Reference Materials) in accordance with ISO 17034 and are measured using validated procedures in our DIN EN ISO/IEC 17025 accredited laboratory. To maintain transparency, we provide a batch-specific certificate of analysis for quality management (QM) documentation.
  • Standards are available in solid and liquid formats. Aquastar® Water Standards in ampoules in different concentrations are for the daily titer determinations and instrument checks. Special standards for the Karl Fischer oven method, an oil standard, as well as a lactose standard for the pharmaceutical sector round off the range.
Modern laboratory titration setup featuring a digital titrator, various transparent reagent bottles with one containing a yellow liquid, and a dark brown bottle with a label, all connected by tubes, alongside a white tray with empty vials, indicating a precise chemical analysis process.
SmartChemicals for Titration and Karl Fischer Titration
Our innovative SmartChemical portfolio enables a connection between the reagents, standards and the titrator. Our Aquastar® Karl FischerTitrants and Aquastar® Water Standards, as well as our Titripur® Volumetric Solutions and Certipur® Volumetric Standards transmit data wirelessly to the titrator. A brief touch on the SmartChemical RFID (Radio Frequency Identification) tag, integrated in the label, eliminates transmission errors, saves time, protects data, and increases efficiency.
Benefits of using SmartChemicals
  • Fast and secure data transfer
  • Improved efficiency
  • Easy to use, easy to implement
  • Extended quality management
Related Resources
Page 1 of 2