Merck
1.17360
Power Supply
new
for use with MAS-100 Sirius®
Note: For More Information on this product, including pricing and availability, please click here!
Passer à
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
Conditionnement
pkg of 1 unit
Fabricant/nom de marque
MAS-100® (Sirius Power Supply )
dilution
(Application - Air monitoring)
Application(s)
pharmaceutical (Comestic, Medical Device; Hospital)
Compatibilité
for use with MAS-100 Sirius®
Description générale
The MAS-100 Sirius® air sampler is equipped with a rechargeable battery pack and USB-C interface for charging.
The power supply is a 36W USB-PD adapter for optimized charging time of the Sirus instrument. The full charging time will be approximately 3.5 hours.
It comes with 4 wall plug in adapters covering US/CA/JP/UK/EU and AU.
Application
The power supply is used to power the MAS-100 Sirius® air sampler that is designed for active air monitoring in cleanrooms of Pharmaceutical, Cosmetic, and Hospital environment.
Informations légales
MAS-100 is a registered trademark of MBV AG
MAS-100 Sirius is a registered trademark of MBV AG
Contenu apparenté
Discover the MAS-100 Sirius® Microbial Air Sampler, designed for reliable air monitoring. Explore features, training, and support services to enhance your lab's compliance and efficiency.