Merck
Passer à
NM0048
NovaSeal Battery Charger
Battery Charger 220 - 240 V UK Plug
Synonyme(s) :
Battery charger for NovaSeal crimping tool
Se connecter pour consulter les tarifs organisationnels et contractuels.
Changer de vue
Taille/RéférenceDisponibilitéPrix
1 each
Vérifier la disponibilité des articles du panier
621,00 €
A propos de cet article
UNSPSC Code:
12352200
eCl@ss:
32039090
621,00 €
Vérifier la disponibilité des articles du panier
​
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
Quality Segment
sterility
non-sterile
product line
NovaSeal
compatibility
for use with NovaSeal battery powered crimping tool - NM0045
Comparer avec des articles similaires
Voir la comparaison complète
Montrer les différences
1 of 1
Cet article
NM0046NM0047NM0042
description
Battery Charger 220 - 240 V UK Plug
description
Battery Charger 100 -120 V US and Japan Plug
description
Battery Charger 220 - 240 V European Plug
description
Battery, 10.8 V Li-ion
compatibility
for use with NovaSeal battery powered crimping tool - NM0045
compatibility
for use with NovaSeal battery powered crimping tool - NM0043
compatibility
for use with NovaSeal battery powered crimping tool - NM0044
compatibility
for use with NovaSeal battery powered crimping tool
Quality Level
100
Quality Level
100
Quality Level
100
Quality Level
-
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
product line
NovaSeal
product line
NovaSeal
product line
NovaSeal
product line
NovaSeal
Still not finding the right product?
Explore all of our products under NovaSeal Battery Charger