Skip to Content
MilliporeSigma

Skip To

Z166340

Quantofix® test strips

Ascorbic acid (Vit.C)

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NB.35
UNSPSC Code:
41113034
eCl@ss:
32129001


description

range mg/L 0, 50, 100, 200, 300, 500, 1000, 2000, semi-quantitative

specific analyte(s)

ascorbic acid

packaging

pack of 100 ea

manufacturer/tradename

Macherey-Nagel

color

yellow to green-blue change

detection method

colorimetric

General description

For semi-quantitative determination of ions and other substances. Consists of plastic strip to which test paper has been sealed. Size of test paper area permits clear comparison with fields of equal size on color scale provided on plastic box. Length sufficient to permit easy handling, even when aggressive or otherwise noxious solutions are being tested.

  • Fast; dip and read
  • No apparatus needed, only single test stick for analysis
  • Highly specific; interferences can be eliminated by precipitation or masking
  • Dim.: 6 × 95 × 0.2 mm

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters