Skip to Content
MilliporeSigma

AOBH97F101E5

AOBChem

2-fluoro-6-formyl-3-methoxybenzoic acid

Synonym(s):

2-fluoro-6-formyl-3-methoxybenzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Product AOBH97F101E5 is not currently sold in your country.

About This Item

CAS Number:
Molecular Weight:
198.15
MDL number:
Product AOBH97F101E5 is not currently sold in your country.

Inchi Code

1S/C9H7FO4/c1-14-6-3-2-5(4-11)7(8(6)10)9(12)13/h2-4H,1H3,(H,12,13)

Inchi Key

UFVQFRBXXKZMEX-UHFFFAOYSA-N

Purity

97%

Country of Origin

CN

Physical Form

Solid

MSDS

https://aobchem.com/2-fluoro-6-formyl-3-methoxybenzoic-acid.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library