Skip to Content
MilliporeSigma

Skip To

BDPH9BCC400A

Bide Pharmatech

Spiro[2.5]octane-6-carbaldehyde

Synonym(s):

spiro[2.5]octane-6-carbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C9H14O1
CAS Number:
Molecular Weight:
138.21
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H14O/c10-7-8-1-3-9(4-2-8)5-6-9/h7-8H,1-6H2

Inchi Key

LQPGRYWUUGVTPV-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Storage temp.

Inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under Spiro[2.5]octane-6-carbaldehyde



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library