Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

BDPH9BCCC19D

Bide Pharmatech

1,4-Dibromo-2,5-dimethylbenzene

Synonym(s):

1,4-dibromo-2,5-dimethylbenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9BCCC19D is not currently sold in your country.

About This Item

Linear Formula:
C8H8Br2
CAS Number:
Molecular Weight:
263.96
MDL number:
Product BDPH9BCCC19D is not currently sold in your country.

Inchi Code

1S/C8H8Br2/c1-5-3-8(10)6(2)4-7(5)9/h3-4H,1-2H3

Inchi Key

QENIALCDPFDFHX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Powder or crystals

Melting Point

72-74C

Looking for similar products? Visit Product Comparison Guide

hstatements

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library