Skip to Content
MilliporeSigma

BDPH9BCD46BB

Bide Pharmatech

2-(2-Chlorophenyl)acetaldehyde

Synonym(s):

(2-chlorophenyl)acetaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H7O1Cl1
CAS Number:
Molecular Weight:
154.6
MDL number:
Product BDPH9BCD46BB is not currently sold in your country.


Inchi Code

1S/C8H7ClO/c9-8-4-2-1-3-7(8)5-6-10/h1-4,6H,5H2

Inchi Key

NTLKDYQFUMXRNF-UHFFFAOYSA-N

Shipping Temperature

Shipped in cold pack

Purity

98%

Country of Origin

CN

Physical Form

Liquid

Storage temp.

Inert atmosphere, store in freezer, under -20°C



Still not finding the right product?

Explore all of our products under 2-(2-Chlorophenyl)acetaldehyde


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library