Skip to Content
MilliporeSigma

Skip To

BDPH9BCD74CA

Bide Pharmatech

7-Bromo-4-methyl-3,4-dihydro-2H-benzo[b][1,4]oxazine

Synonym(s):

7-bromo-4-methyl-3,4-dihydro-2H-1,4-benzoxazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C9H10O1N1Br1
CAS Number:
Molecular Weight:
228.09
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H10BrNO/c1-11-4-5-12-9-6-7(10)2-3-8(9)11/h2-3,6H,4-5H2,1H3

Inchi Key

MQMFOFZKZBLSAB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Light green or light yellow to Brown liquid or solid



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library