Skip to Content
MilliporeSigma

Skip To

BDPH9BCD8A65

Bide Pharmatech

3-Aminopentanoic acid

Synonym(s):

3-aminopentanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C5H11O2N1
CAS Number:
Molecular Weight:
117.15
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H11NO2/c1-2-4(6)3-5(7)8/h4H,2-3,6H2,1H3,(H,7,8)

Inchi Key

QFRURJKLPJVRQY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Colorless to White to Yellow Solid or Liquid



Still not finding the right product?

Explore all of our products under 3-Aminopentanoic acid


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library