Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C15H12Br2O/c16-13-5-1-11(2-6-13)9-15(18)10-12-3-7-14(17)8-4-12/h1-8H,9-10H2
Inchi Key
DQQKEYDDVSREIE-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 1,3-Bis(4-bromophenyl)propan-2-one
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
