Skip to Content
MilliporeSigma

BDPH9BCDC1CE

Bide Pharmatech

1-(Quinolin-8-yl)ethanone

Synonym(s):

1-(8-quinolinyl)ethanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H9O1N1
CAS Number:
Molecular Weight:
171.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H9NO/c1-8(13)10-6-2-4-9-5-3-7-12-11(9)10/h2-7H,1H3

Inchi Key

SHVCKCOMBWIPBE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Yellow to Brown Solid



Still not finding the right product?

Explore all of our products under 1-(Quinolin-8-yl)ethanone


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library