Skip to Content
MilliporeSigma

Skip To

BDPH9BCDDE9F

Bide Pharmatech

2-(2-Chlorophenyl)cyclopentan-1-amine

Synonym(s):

2-(2-chlorophenyl)cyclopentanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C11H14N1Cl1
CAS Number:
Molecular Weight:
195.69
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H14ClN/c12-10-6-2-1-4-8(10)9-5-3-7-11(9)13/h1-2,4,6,9,11H,3,5,7,13H2

Inchi Key

GNPPIVFFQGMMCI-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library