Skip to Content
MilliporeSigma

BDPH9BCDF3E6

Bide Pharmatech

3-(tert-Butyl)-1-phenyl-1H-pyrazole-5-carboxylic acid

Synonym(s):

3-tert-butyl-1-phenyl-1H-pyrazole-5-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C14H16O2N2
CAS Number:
Molecular Weight:
244.29
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C14H16N2O2/c1-14(2,3)12-9-11(13(17)18)16(15-12)10-7-5-4-6-8-10/h4-9H,1-3H3,(H,17,18)

Inchi Key

JYAMNOJMPNSKGL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library