Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day through March 27.Save Now

BDPH9BCDFD2D

Bide Pharmatech

1-(((2R,3R,4R,5R,6R)-3-Acetamido-4,5-diacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-16,16-bis((3-((3-(5-(((2R,3R,4R,5R,6R)-3-acetamido-4,5-diacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)oxy)pentanamido)propyl)amino)-3-oxopropoxy)methyl)-5,11,18-trioxo-14-oxa-6,10,17-triazanonacosan-29-oic acid

Synonym(s):

1-(((2R,3R,4R,5R,6R)-4,5-diacetoxy-6-(acetoxymethyl)-3-((1-hydroxyethylidene)amino)tetrahydro-2H-pyran-2-yl)oxy)-16,16-bis((3-((3-((5-(((2R,3R,4R,5R,6R)-4,5-diacetoxy-6-(acetoxymethyl)-3-((1-hydroxyethylidene)amino)tetrahydro-2H-pyran-2-yl)oxy)-1-hydroxypentylidene)amino)propyl)imino)-3-hydroxypropoxy)methyl)-5,11,18-trihydroxy-14-oxa-6,10,17-triazanonacosa-5,10,17-trien-29-oic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9BCDFD2D is not currently sold in your country.

Product BDPH9BCDFD2D is not currently sold in your country.

Inchi Code

1S/C91H148N10O39/c1-56(102)98-79-85(135-65(10)111)82(132-62(7)108)68(50-129-59(4)105)138-88(79)126-44-24-21-30-71(114)92-38-27-41-95-74(117)35-47-123-53-91(101-77(120)33-19-17-15-13-14-16-18-20-34-78(121)122,54-124-48-36-75(118)96-42-28-39-93-72(115)31-22-25-45-127-89-80(99-57(2)103)86(136-66(11)112)83(133-63(8)109)69(139-89)51-130-60(5)106)55-125-49-37-76(119)97-43-29-40-94-73(116)32-23-26-46-128-90-81(100-58(3)104)87(137-67(12)113)84(134-64(9)110)70(140-90)52-131-61(6)107/h68-70,79-90H,13-55H2,1-12H3,(H,92,114)(H,93,115)(H,94,116)(H,95,117)(H,96,118)(H,97,119)(H,98,102)(H,99,103)(H,100,104)(H,101,120)(H,121,122)/t68-,69-,70-,79-,80-,81-,82+,83+,84+,85-,86-,87-,88-,89-,90-/m1/s1

Inchi Key

DHFOAYZFYXIPOR-CIGQGGIZSA-N

Shipping Temperature

Cold-chain transportation

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Powder or crystals

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library