Skip to Content
MilliporeSigma

Skip To

BDPH9C01EC7D

Bide Pharmatech

PSMA-ALB-56

Synonym(s):

(3S,10S,14S)-1,4,12-trihydroxy-1-(4-((((S)-1-hydroxy-2-((1-hydroxy-2-(4,7,10-tris(carboxymethyl)-1,4,7,10-tetraazacyclododecan-1-yl)ethylidene)amino)-6-((1-hydroxy-4-(p-tolyl)butylidene)amino)hexylidene)amino)methyl)cyclohexyl)-3-(naphthalen-2-ylmethyl)-2,5,11,13-tetraazahexadeca-1,4,11-triene-10,14,16-tricarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C66H95N11O18/c1-44-15-17-45(18-16-44)9-8-14-55(78)67-27-6-4-12-51(70-56(79)40-74-29-31-75(41-58(82)83)33-35-77(43-60(86)87)36-34-76(32-30-74)42-59(84)85)62(89)69-39-46-19-23-49(24-20-46)61(88)71-54(38-47-21-22-48-10-2-3-11-50(48)37-47)63(90)68-28-7-5-13-52(64(91)92)72-66(95)73-53(65(93)94)25-26-57(80)81/h2-3,10-11,15-18,21-22,37,46,49,51-54H,4-9,12-14,19-20,23-36,38-43H2,1H3,(H,67,78)(H,68,90)(H,69,89)(H,70,79)(H,71,88)(H,80,81)(H,82,83)(H,84,85)(H,86,87)(H,91,92)(H,93,94)(H2,72,73,95)/t46-,49-,51-,52-,53-,54-/m0/s1

Inchi Key

QIWYPMRCMDYQBF-HNLNAKQBSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under PSMA-ALB-56




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library