Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

BDPH9C02027C

Bide Pharmatech

N-((S)-5-(((1R,2S)-2-(4-Fluorophenyl)cyclopropyl)amino)-1-(4-methylpiperazin-1-yl)-1-oxopentan-2-yl)-4-(1H-1,2,3-triazol-1-yl)benzamide bis(4-methylbenzenesulfonate)

Synonym(s):

N-((S)-5-(((1R,2S)-2-(4-fluorophenyl)cyclopropyl)amino)-1-(4-methylpiperazin-1-yl)-1-oxopentan-2-yl)-4-(1H-1,2,3-triazol-1-yl)benzamide bis(4-methylbenzenesulfonate)

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9C02027C is not currently sold in your country.

About This Item

Linear Formula:
C42H50O8N7S2F1
CAS Number:
Molecular Weight:
864.03
MDL number:
Product BDPH9C02027C is not currently sold in your country.

Inchi Code

1S/C28H34FN7O2.2C7H8O3S/c1-34-15-17-35(18-16-34)28(38)25(3-2-12-30-26-19-24(26)20-4-8-22(29)9-5-20)32-27(37)21-6-10-23(11-7-21)36-14-13-31-33-36;2*1-6-2-4-7(5-3-6)11(8,9)10/h4-11,13-14,24-26,30H,2-3,12,15-19H2,1H3,(H,32,37);2*2-5H,1H3,(H,8,9,10)/t24-,25-,26+;;/m0../s1

Inchi Key

WLRWBXFBIJMKHG-DVNXTAPYSA-N

Shipping Temperature

Normal

Purity

99%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals

Looking for similar products? Visit Product Comparison Guide

hstatements

signalword

Warning

pictograms

Health hazard

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library