Skip to Content
MilliporeSigma

Skip To

BDPH9C021A4B

Bide Pharmatech

3-Amino-2,4,5-trifluorobenzoic acid

Synonym(s):

3-amino-2,4,5-trifluorobenzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C7H4O2N1F3
CAS Number:
Molecular Weight:
191.11
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Storage temp.

Keep in dark place, inert atmosphere, 2-8°C

Inchi Code

1S/C7H4F3NO2/c8-3-1-2(7(12)13)4(9)6(11)5(3)10/h1H,11H2,(H,12,13)

Inchi Key

NVWVZPFPZJYLNK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library