Skip to Content
MilliporeSigma

BDPH9C021B4E

Bide Pharmatech

(3S,4S)-1-Benzylpyrrolidine-3,4-diyl bis(4-methylbenzenesulfonate)

Synonym(s):

(3S,4S)-1-benzylpyrrolidine-3,4-diyl bis(4-methylbenzenesulfonate)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Product BDPH9C021B4E is not currently sold in your country.


Inchi Code

1S/C25H27NO6S2/c1-19-8-12-22(13-9-19)33(27,28)31-24-17-26(16-21-6-4-3-5-7-21)18-25(24)32-34(29,30)23-14-10-20(2)11-15-23/h3-15,24-25H,16-18H2,1-2H3/t24-,25-/m0/s1

Inchi Key

AIFJHMSGSPJHOH-DQEYMECFSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library