Skip to Content
MilliporeSigma

Skip To

BDPH9C021DBF

Bide Pharmatech

Boc-D-Tryptophanol

Synonym(s):

tert-butyl (1R)-2-hydroxy-1-(1H-indol-3-ylmethyl)ethylcarbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C16H22O3N2
CAS Number:
Molecular Weight:
290.36
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C16H22N2O3/c1-16(2,3)21-15(20)18-12(10-19)8-11-9-17-14-7-5-4-6-13(11)14/h4-7,9,12,17,19H,8,10H2,1-3H3,(H,18,20)/t12-/m1/s1

Inchi Key

JEFQUFUAEKORKL-GFCCVEGCSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

White to light yellow powder or crystals



Still not finding the right product?

Explore all of our products under Boc-D-Tryptophanol


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library