Skip to Content
MilliporeSigma

Skip To

COM448635575

Combi-Blocks, Inc.

1-(5-Bromopyridin-2-yl)azepane

Synonym(s):

1-(5-bromo-2-pyridinyl)azepane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C11H15N2Br1
CAS Number:
Molecular Weight:
255.16
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H15BrN2/c12-10-5-6-11(13-9-10)14-7-3-1-2-4-8-14/h5-6,9H,1-4,7-8H2

Inchi Key

MTYJNKMYXIMVFL-UHFFFAOYSA-N

Purity

97%

Storage temp.

Refrigerated

Country of Origin

CN



Still not finding the right product?

Explore all of our products under 1-(5-Bromopyridin-2-yl)azepane




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library