About This Item
Skip To
packaging
pkg of 1 unit
manufacturer/tradename
MAS-100® (Sirius Power Supply )
dilution
(Application - Air monitoring)
application(s)
pharmaceutical (Comestic, Medical Device; Hospital)
compatibility
for use with MAS-100 Sirius®
General description
The power supply is a 36W USB-PD adapter for optimized charging time of the Sirus instrument. The full charging time will be approximately 3.5 hours.
It comes with 4 wall plug in adapters covering US/CA/JP/UK/EU and AU.
Application
Legal Information
related product
Related Content
Discover the MAS-100 Sirius® Microbial Air Sampler, designed for reliable air monitoring. Explore features, training, and support services to enhance your lab's compliance and efficiency.