MilliporeSigma
1.17653
Wi-Fi Dongle
new
for use with MAS-100® Sirius
Sign In to View Organizational & Contract Pricing
Select a Size
Pricing and availability is not currently available.
Skip To
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pkg of 1 unit
manufacturer/tradename
(Wi-Fi Dongle Sirius)
dilution
(Application - Air monitoring)
application(s)
pharmaceutical (Cosmetic, Medical Device; Hospital)
compatibility
for use with MAS-100® Sirius
General description
The Wi-Fi Dongle for MAS-100 Sirius is designed to allow various wireless operations for the air sampler such as:
  • Automated date/time synchronization
  • Automated timer calibration
  • Automated user synchronization
  • Active Directory login
  • Wireless access to the browser-based user interface (e.g. audit trail review)
Remote control
Application
The Wi-Fi Dongle can be conveniently placed beneath the button plate of the MAS-100 Sirius. Once installed, the instrument seamlessly integrates into your local Wi-Fi network.
Legal Information
MAS-100 is a registered trademark of MBV AG