MilliporeSigma
1.17655
Millipore
HEPA Exhaust filter
new
for use with MAS-100® Sirius®
Slide 1 of 2
Note: For More Information on this product, including pricing and availability, please click here!
Skip To
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
(Polypropylene)
packaging
pkg of 1 unit
manufacturer/tradename
MAS-100® HEPA Exhaust Filter
dilution
(Application - Air monitoring)
compatibility
for use with MAS-100® Sirius®
General description
The MAS-100 Sirius® air sampler features a HEPA exhaust filter classified as an ISO 35 H (H13) filter. This filter is constructed from polypropylene and is embedded in a frame made of polyethylene terephthalate (PET). Its primary function is to prevent the release of contaminants into the cleanroom during the sampling process, particularly when the instrument is transferred from lower to higher-grade cleanrooms.
The Sirius supported by the efficient exhaust HEPA filter operates effectively in both non-controlled areas and cleanroom environments up to grade A (or ISO 5), maintaining compliance with viable and non-viable particle limits. The Exhaust filter is mounted directly at the exhaust of the air path of the MAS-100 Sirius® air sampler. It is recommended to replace the filter on a yearly basis, e.g. during calibration of the air sampler. It can easily be exchanged without the need of any tools.
Application
HEPA exhaust filter in the MAS-100 Sirius® air sampler is essential for maintaining contamination control and high air quality standards in cleanroom environments. It prevents the release of airborne contaminants during the sampling process, ensuring the integrity of the cleanroom is preserved and aiding compliance with stringent regulatory requirements in pharmaceutical, cosmetic, biotechnology, and medical settings.
Legal Information
MAS-100 is a registered trademark of MBV AG
MAS-100 Sirius is a registered trademark of MBV AG
Visit the Document Library
Related Content
Discover the MAS-100 Sirius® Microbial Air Sampler, designed for reliable air monitoring. Explore features, training, and support services to enhance your lab's compliance and efficiency.