Skip to Content
MilliporeSigma

ABE1788

Anti-Nicotinamide N-methyltransferase

from rabbit, purified by affinity chromatography

Synonym(s):

EC 2.1.1.1

Sign In to View Organizational & Contract Pricing

Select a Size

25 μG

$247.00

100 μG

$623.00

$247.00


Please contact Customer Service for Availability

Request a Bulk Order

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... NNMT(4837)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB1402290SAB1410844MABS1822
antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

mouse

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

shipped in

ambient

shipped in

dry ice

shipped in

dry ice

shipped in

ambient

clone

polyclonal

clone

2F2, monoclonal

clone

polyclonal

clone

4D2.1, monoclonal

Gene Information

human ... NNMT(4837)

Gene Information

human ... NNMT(4837)

Gene Information

human ... NNMT(4837)

Gene Information

human ... SHMT2(6472)

General description

Nicotinamide N-methyltransferase (UniProt: P40261; also known as EC: 2.1.1.1) is encoded by the NNMT gene (Gene ID: 4837) in human. Nicotinamide N-methyltransferase is a monomeric, cytoplasmic enzyme that belong to the class I -like SAM-binding methyltransferase superfamily. It catalyzes the N-methylation of nicotinamide and other pyridines with S-Adenosyl-L-methionine (SAM)) serving as the methyl donor. It has two SAM binding sites (aa 63-64 and 142-143). Its catalytic activity is important for biotransformation of many drugs and xenobiotic compounds. Nicotinamide N-methyltransferase is predominantly expressed in the liver. A lower level of expression is seen in the kidney, lung, skeletal muscle, placenta, and heart and it is not detected in the brain or pancreas. Nicotinamide N-methyltransferase inhibition is shown to increases adipose SAM and NAD(+) levels and upregulate ornithine decarboxylase and spermidine-spermine N(1)-acetyltransferase activity and expression. (Ref.: Kraus, D et al. (2014). Nature. 508 (7495); 258-62).
~30 kDa observed; 29.57 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the internal region of human Nicotinamide N-methyltransferase.

Application

Anti-Nicotinamide N-methyltransferase Antibody, Cat. No. ABE1788, is a highly specific rabbit polyclonal antibody that targets human Nicotinamide N-methyltransferase and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: 1 µg/mL from a representative lot detected Nicotinamide N-methyltransferase in 10 µg of human kidney lysate and HEK293 cell lysate.

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Nicotinamide N-methyltransferase in human liver, human kidney, and human pancreas tissues.

Biochem/physiol Actions

This rabbit polyclonal antibody detects human Nicotinamide N-methyltransferase. It targets an epitope within 15 amino acids from the internal region.

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Nicotinamide N-methyltransferase in 10 µg of HepG2 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service