Skip to Content
MilliporeSigma

Skip To

WISH (HeLa derivative) Human Cervix, ECACC 88102403

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41106514
Biological source:
human cervix
Relevant disease(s):
cancer
Growth mode:
Adherent
Karyotype:
2n = 46
Morphology:
Epithelial
Manufacturer/tradename:
ECACC
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

WISH (HeLa derivative), NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

human cervix

manufacturer/tradename

ECACC

packaging

tube of 5 μg 88102403-DNA-5UG, pkg of vial of cells 88102403-1VL

growth mode

Adherent

karyotype

2n = 46

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Application

Virus studies: VSV (Indiana), Ad 3, Poliovirus, measles virus

Biochem/physiol Actions

Human cervix carcinoma
Originally derived from human amnion tissue. Have been used in virus studies; susceptible to VSV (Indiana), adenovirus 3 and poliovirus. Useful in differentiating virulent and avirulent measles virus. HeLa marker chromosomes and type A G6PD have been detected. This cell line was found to be indistinguishable from HeLa by STR PCR DNA profiling. Therefore, the cell line must be considered as derived from HeLa. Ethnicity: Black.
STR-PCR Data: Amelogenin: X
CSF1PO: 9,10
D13S317: 14
D16S539: 9,10
D5S818: 11,12
D7S820: 12
THO1: 7
TPOX: 8,12
vWA: 16,18

Preparation Note

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 15% Foetal Bovine Serum (FBS).
Split sub-confluent cultures (70-80%) 1:2 to 1:10 i.e. seeding at 1-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CB_90111910CB_93120839CB_93120824
description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

human cervix

biological source

human cervix

biological source

human cervix

biological source

human cervix

products

Not specified

products

Not specified

products

Not specified

products

-

growth mode

Adherent

growth mode

Adherent

growth mode

Adherent

growth mode

-

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

-

receptors

Not specified

receptors

Not specified

receptors

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters