Skip to Content
MilliporeSigma

F8318

Fetal Bovine Serum

Australia origin, sterile-filtered, suitable for cell culture, suitable for hybridoma

Synonym(s):

Fetal Bovine Serum, Fetal Calf Serum, FBS, FCS, sera, serum

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
12352207
MDL number:
Biological source:
bovine fetus
Origin:
Australia origin
Sterility:
sterile-filtered
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Fetal Bovine Serum, Australia origin, sterile-filtered, suitable for cell culture, suitable for hybridoma

biological source

bovine fetus

sterility

sterile-filtered

product line

FBS Classic

origin

Australia origin

technique(s)

cell culture | hybridoma: suitable
cell culture | mammalian: suitable

storage temp.

−20°C

Quality Level

Looking for similar products? Visit Product Comparison Guide

Analysis Note

  • Endotoxin and hemoglobin tested
Endotoxin and hemoglobin tested

General description

Animal serum is commonly used to supplement basal media formulations for the optimal growth of many cell types in vitro.
Fetal Bovine Serum (FBS) is the most common serum used to supplement cell culture media due to its high nutritional content. Although it is relatively low in protein, FBS is effective in promoting and sustaining growth of vertebrate mammalian and insect cells

Preparation Note

To effectively preserve the integrity of animal serum, it should be stored frozen and protected from light. Multiple thaw/freeze cycles should be avoided as they will hasten the degradation of serum nutrients and can result in the formation of insoluble precipitates.
For stability and optimal performance, serum should be stored at −20 °C and used prior to the labeled expiration date.

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Découvrez notre gamme de sérums de veau fœtal (FBS) et identifiez le FBS adapté à vos besoins et à votre budget. Demander un échantillon de sérum.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service