Skip to Content
MilliporeSigma

O7251

Oleic anhydride

~99%, liquid

Synonym(s):

cis-9-Octadecenoic anhydride

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
[CH3(CH2)7CH=CH(CH2)7CO]2O
CAS Number:
Molecular Weight:
546.91
EC Number:
246-522-4
UNSPSC Code:
12352211
MDL number:
Beilstein/REAXYS Number:
1730211
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/C36H66O3/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35(37)39-36(38)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20H,3-16,21-34H2,1-2H3/b19-17-,20-18-

InChI key

OCNZHGHKKQOQCZ-CLFAGFIQSA-N

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OC(=O)CCCCCCC\C=C/CCCCCCCC

assay

~99%

form

liquid

bp

200-215 °C/11 mmHg (lit.)

mp

22-24 °C (lit.)

storage temp.

−20°C

Packaging

Sealed ampule.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

228.2 °F - closed cup

flash_point_c

109 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service