Skip to Content
MilliporeSigma

Skip To

P1498

Anti-Heat Shock Protein 27 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-HSP27

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ARR, IF, WB
Citations:
9


biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 27 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: 1:250-1:500 using HeLa cells, microarray: suitable, western blot: 1:4,000-1:8,000 using whole cell extract of human epitheloid carcinoma HeLa cell line or human epidermoid carcinoma A431 cell line

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSPB1(3315)
mouse ... Hspb1(15507)
rat ... Hspb1(24471)

General description

Heat shock protein 27 (HSP27) is an actin-binding protein, that is located on human chromosome 7q11.23. HSP27 (mouse HSP25) is a highly conserved oligomeric protein. This protein is expressed in various cell types and tissues during specific development and differentiation stages.

Immunogen

synthetic peptide corres­ponding to amino acids 186-205 located at the C-terminus of human HSP27, conjugated to KLH. This sequence is highly homologous in rat HSP27 (85% identity) and to a lesser extent in mouse HSP27/25 (65% identity).

Application

Anti-Heat Shock Protein 27 antibody produced in rabbit may be used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Anti- Heat Shock Protein 27 (HSP27) recognizes HSP27 (27 kDa).
Heat shock protein 27 (HSP27) accumulates in cells exposed to a short period of hyperthermia and contributes to the development of a transient state of thermotolerance. HSP27 can inhibit actin polymerization, thus modulating actin dynamics during stress. It is known to guard against apoptotic cell death activated by several stimuli, such as Fas ligand hyperthermia, oxidative stress and cytotoxic drugs. It can hinder the mitochondrial pathway of caspase-induced cell death by acting as a negative regulator of cytochrome c-dependent activation of caspase-3. The synthesis of HSP27 is stimulated by various cytokines, growth factors, hormones and chemicals. HSP27 shows a rapid phosphorylation, following exposure to stress stimuli. HSP27 is expressed in the cytoplasm and colocalizes to the nucleus upon stress stimuli. Mutation on this HSP27 gene is associated with Charcot-Marie-Tooth disease.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

For continuous use, store at 2-8°C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters