Skip to Content
MilliporeSigma

78687

α-Aminoisobutyric acid

new

analytical standard

Synonym(s):

2-Methylalanine, Aib

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Linear Formula:
(CH3)2C(NH2)COOH
CAS Number:
Molecular Weight:
103.12
Beilstein:
506496
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
LC/MS: suitable
liquid chromatography (LC): suitable
mass spectrometry (MS): suitable

mp

≥300 °C

application(s)

clinical
food and beverages

format

neat

SMILES string

CC(C)(N)C(O)=O

InChI

1S/C4H9NO2/c1-4(2,5)3(6)7/h5H2,1-2H3,(H,6,7)

InChI key

FUOOLUPWFVMBKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Aminoisobutyric acid (AIB; 2‑aminoisobutyric acid, α‑methylalanine) is a non‑proteinogenic amino acid with the formula H2N‑C(CH3)2‑COOH that strongly promotes helical conformations in peptides.

It is rare in nature but occurs in certain fungal peptide antibiotics (peptaibols) such as alamethicin, where AIB-rich sequences contribute to membrane activity.Testing AIB is important because it is used as a tracer of amino acid transport and as a sensitive marker of blood–brain barrier permeability changes in experimental models.

In metabolic studies, AIB can be measured in urine as an end‑product of pyrimidine metabolism, and some individuals exhibit elevated excretion due to inherited traits, making quantification relevant for biochemical assessment. Additionally, detecting AIB helps identify peptaibiotics in microbial or environmental samples, since AIB is a specific marker residue for this peptide class.

Discover our portfolio of amino acid reference materials

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service