MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • A physics-based temperature model for ultrasonic vibration-assisted pelleting of cellulosic biomass.
A physics-based temperature model for ultrasonic vibration-assisted pelleting of cellulosic biomass.
Ultrasonics (2014-06-18)
Xiaoxu Song, Xiaoming Yu, Meng Zhang, Z J Pei, Donghai Wang
ABSTRACT
Temperature in ultrasonic vibration-assisted (UV-A) pelleting of cellulosic biomass has a significant impact on pellet quality. However, there are no reports on temperature models for UV-A pelleting of cellulosic biomass. The development of a physics-based temperature model can help to explain experimentally determined relations between UV-A pelleting process variables and temperature, and provide guidelines to optimize these process variables in order to produce pellets of good quality. This paper presents such a model for UV-A pelleting of cellulosic biomass. Development of the model is described first. Then temperature distribution is investigated using the model, and temperature difference between the top and the bottom surfaces of a pellet is explained. Based on this model, relations between process variables (ultrasonic power and pelleting duration) and temperature are predicted. Experiments were conducted for model verification, and the results agreed well with model predictions.
Product Description
Supelco
Cellulose, acid washed, from spruce, suitable for column chromatography
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Sigma-Aldrich
Cellulose, fibers, (medium)
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, acid washed, powder, suitable for column chromatography
Sigma-Aldrich
Cellulose, microcrystalline, powder
Supelco
Cellulose, powder, suitable for column chromatography
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
α-Cellulose, powder
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers