MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Co-translational localization of an LTR-retrotransposon RNA to the endoplasmic reticulum nucleates virus-like particle assembly sites.
Co-translational localization of an LTR-retrotransposon RNA to the endoplasmic reticulum nucleates virus-like particle assembly sites.
PLoS genetics (2014-03-08)
Jung H Doh, Sheila Lutz, M Joan Curcio
ABSTRACT
The transcript of retrovirus-like transposons functions as an mRNA for synthesis of capsid and replication proteins and as the genomic RNA of virus-like particles (VLPs), wherein the genome is replicated. Retrotransposon RNA and proteins coalesce in a cytoplasmic focus, or retrosome, to initiate VLP assembly, but it is not known how the retrosome is nucleated. We determined how the RNA and Gag protein of the Saccharomyces cerevisiae Ty1 retrotransposon are directed to the retrosome. We found that Ty1 RNA is translated in association with signal recognition particle (SRP), a universally conserved chaperone that binds specific ribosome-nascent chain (RNC) complexes and targets the nascent peptide to the endoplasmic reticulum (ER). Gag is translocated to the ER lumen; yet, it is also found in the cytoplasm, associated with SRP-RNC complexes. In the absence of ER translocation, Gag is synthesized but rapidly degraded, and Ty1 RNA does not coalesce in retrosomes. These findings suggest that Gag adopts a stable conformation in the ER lumen, is retrotranslocated to the cytoplasm, binds to Ty1 RNA on SRP-RNC complexes and multimerizes to nucleate retrosomes. Consistent with this model, we show that slowing the rate of co-translational ER translocation by limiting SRP increases the prevalence of retrosomes, while suppressing the translocation defect of srp hypomorphs by slowing translational elongation rapidly decreases retrosome formation. Thus, retrosomes are dynamic foci of Ty1 RNA-RNC complexes whose formation is modulated by the rate of co-translational ER translocation. Together, these findings suggest that translating Ty1 mRNA and the genomic RNA of VLPs originate in a single pool and moreover, that co-translational localization of Ty1 RNA nucleates the presumptive VLP assembly site. The separation of nascent Gag from its RNA template by transit through the ER allows Gag to bind translating Ty1 RNA without displaying a cis-preference for its encoding RNA.
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
3-Methyl-1-butanol, ACS reagent, ≥98.5%
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%
Sigma-Aldrich
3-Methyl-1-butanol, reagent grade, 98%
Supelco
3-Methyl-1-butanol, analytical standard
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Supelco
3-Methylbutanol, analytical standard
Sigma-Aldrich
1H-1,2,3-Triazole, 97%
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Isoamyl alcohol, ≥98%, FG
Sigma-Aldrich
Isoamyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
3-Methylbutanol, BioUltra, Molecular Biology, ≥99.0% (GC)