MilliporeSigma
  • Long-range evolutionary constraints reveal cis-regulatory interactions on the human X chromosome.
Long-range evolutionary constraints reveal cis-regulatory interactions on the human X chromosome.
Nature communications (2015-04-25)
Magali Naville, Minaka Ishibashi, Marco Ferg, Hemant Bengani, Silke Rinkwitz, Monika Krecsmarik, Thomas A Hawkins, Stephen W Wilson, Elizabeth Manning, Chandra S R Chilamakuri, David I Wilson, Alexandra Louis, F Lucy Raymond, Sepand Rastegar, Uwe Strähle, Boris Lenhard, Laure Bally-Cuif, Veronica van Heyningen, David R FitzPatrick, Thomas S Becker, Hugues Roest Crollius
ABSTRACT
Enhancers can regulate the transcription of genes over long genomic distances. This is thought to lead to selection against genomic rearrangements within such regions that may disrupt this functional linkage. Here we test this concept experimentally using the human X chromosome. We describe a scoring method to identify evolutionary maintenance of linkage between conserved noncoding elements and neighbouring genes. Chromatin marks associated with enhancer function are strongly correlated with this linkage score. We test >1,000 putative enhancers by transgenesis assays in zebrafish to ascertain the identity of the target gene. The majority of active enhancers drive a transgenic expression in a pattern consistent with the known expression of a linked gene. These results show that evolutionary maintenance of linkage is a reliable predictor of an enhancer's function, and provide new information to discover the genetic basis of diseases caused by the mis-regulation of gene expression.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Ethyl 3-aminobenzoate methanesulfonate, 98%
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)