MilliporeSigma
HPLC Analysis of Spironolactone on Chromolith® HighResolution RP-18 endcapped 100 x 4.6 mm
HPLC Analysis of Spironolactone on Chromolith® HighResolution RP-18 endcapped 100 x 4.6 mm application for HPLC
CONDITIONS
column
Chromolith® HighResolution RP-18 endcapped, 100 x 4.6 mm (1.52022.0001)
mobile phase
[A] water; [B] methanol; (40:60, A:B)
flow rate
1.5 mL/min
pressure
1175 psi (81 bar)
column temp.
ambient
detector
230 nm (cell: 11 μL)
injection
13 μL (reduced with 33% as it is scaled to a 33% shorter column with same diameter)
sample
0.5 mg/mL (500 ppm) of Spironolactone RS in diluent (diluent: acetonitrile/water 1:1 (v/v))
Description
Legal Information
Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany