MilliporeSigma
HPLC Analysis of Spironolactone on Chromolith® HighResolution RP-18 endcapped 50 x 4.6 mm
HPLC Analysis of Spironolactone on Chromolith® HighResolution RP-18 endcapped 50 x 4.6 mm application for HPLC
CONDITIONS
column
Chromolith® HighResolution RP-18 endcapped, 50 x 4.6 mm (1.52021.0001)
mobile phase
[A] water; [B] methanol; (40:60,:A:B)
flow rate
1.5 mL/min
pressure
856 psi (59 bar)
column temp.
ambient
detector
230 nm cell: 11μL
injection
7 μL (reduced with 67% as it is scaled to a 67% shorter column with same diameter)
sample
0.5 mg/mL, 500 ppm Spironolactone RS in diluent (diluent: acetonitrile/water 1:1 (v/v))
Description
Legal Information
Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany