MilliporeSigma
HomeReaction Design & OptimizationAsymmetric Reduction of Ketones
Asymmetric Reduction of Ketones
The enantioselective reduction of ketones to alcohols gives access to a pool of chiral building blocks that can be used for the synthesis of natural products. Adolfsson and co-workers reported a novel class of ligands, based on a pseudo-dipeptide,1 for the mild and efficient reduction of ketones using 2-propanol as a hydrogen source. The ligand is used with [{RuCl2(p-cymene)}2] in presence of NaOH for the asymmetric reduction of a variety of acetophenone derivatives. Excellent yields and ee’s were reported (Scheme 1).
Asymmetric Reduction of Ketones
Scheme 1.
Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
References
1.
Bøgevig A, Pastor IM, Adolfsson H. 2004. Highly Enantioselective Ruthenium-Catalyzed Reduction of Ketones Employing Readily Available Peptide Ligands. Chem. Eur. J.. 10(1):294-302. https://doi.org/10.1002/chem.200305553