Merck
  • Reciprocal feedback inhibition of the androgen receptor and PI3K as a novel therapy for castrate-sensitive and -resistant prostate cancer.
Reciprocal feedback inhibition of the androgen receptor and PI3K as a novel therapy for castrate-sensitive and -resistant prostate cancer.
Oncotarget (2015-10-28)
Wenqing Qi, Carla Morales, Laurence S Cooke, Benny Johnson, Bradley Somer, Daruka Mahadevan
ABSTRACT
Gain-of-function of the androgen receptor (AR) and activation of PI3K/AKT/mTOR pathway have been demonstrated to correlate with progression to castration-resistant prostate cancer (CRPC). However, inhibition of AR or PI3K/mTOR alone results in a reciprocal feedback activation. Therefore, we hypothesized that dual inhibition of the AR and PI3K/mTOR pathway might lead to a synergistic inhibition of cell growth and overcome drug resistance in CRPC. Here, we reported that androgen-depletion increased AR protein level and Akt phosphorylation at Ser473 and Thr308 in LNCaP cells. Moreover, we developed resistance cell lines of LNCaP to Enzalutamide (or MDV3100), an AR inhibitor (named as LNCaP 'MDV-R') and PF-04691502, a PI3K/mTOR inhibitor (named as LNCaP 'PF-R'). MTS analysis showed that LNCaP 'PF-R' was strongly resistant to Enzalutamide treatment, and on the other hand, LNCaP 'MDV-R' was 6-fold resistant to PF-04691502 treatment. Mechanistically, LNCaP 'MDV-R' cells had significantly reduced AR, loss of PSA and increase Akt activity in contrast with LNCaP 'PF-R' cells. Combined inhibition of PI3K/mTOR and AR pathways with a variety of small molecular inhibitors led to a synergistic suppression of cell proliferation and a profound increase of apoptosis and cell cycle arrest in both androgen-dependent LNCaP and independent CRPC 22Rv1 cell lines. In conclusion, this study provides preclinical proof-of-concept that the combination of a PI3K/mTOR inhibitor with an AR inhibitor results in a synergistic anti-tumor response in non-CRPC and CRPC models.
Product Description
Supelco
DL-Dithiothreitol solution, 1 M in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)