Merck
BMSBSH50022BEUK
myBlock ll digital dry bath with 2 Quick-Flip blocks for tube
strips and plates, AC/DC input 230 V AC, UK plug
Se connecter pour consulter les tarifs organisationnels et contractuels.
Le tarif et la disponibilité ne sont pas disponibles actuellement.
A propos de cet article
NACRES:
NB.76
UNSPSC Code:
41102404
Passer à
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
Nom du produit
myBlock ll digital dry bath with 2 Quick-Flip blocks for tube, strips and plates, AC/DC input 230 V AC, UK plug
manufacturer/tradename
Benchmark Scientific BSH5002-2B-E-UK
parameter
+5 °C temp. range (+105 °C)
5-105 °C temp. range
capacity
, holds 2 blocks
Comparer avec des articles similaires
Voir la comparaison complète
Montrer les différences
1 of 4
Cet article
BMSBSH5002EUKBMSBSH50011BEUKBMSBSH5001EUK
capacity
, holds 2 blocks
capacity
, holds 2 blocks
capacity
, holds 1 block
capacity
, holds 1 block
parameter
+5 °C temp. range (+105 °C), 5-105 °C temp. range
parameter
5-105 °C temp. range
parameter
+5 °C temp. range (+105 °C), 1 min to 99 hr 59 min (Timer), 105 °C temp. range, Ambient +5 to 105 °C temp. range
parameter
+5 °C temp. range (+105 °C), 1 min to 99 hr 59 min (Timer), 5-105 °C temp. range, Ambient +5 to 105 °C temp. range
manufacturer/tradename
Benchmark Scientific BSH5002-2B-E-UK
manufacturer/tradename
Benchmark Scientific BSH5002-E-UK
manufacturer/tradename
Benchmark Scientific BSH5001-1B-E-UK
manufacturer/tradename
Benchmark Scientific BSH5001-E-UK
General description
myBlock series is a "next generation" product line that introduces several major advantages over traditional dry baths. They are the first digital dry baths to include advanced microprocessor controls, timed or continuous operation and a removable hinged lid. In addition, an optional external temperature feedback probe can be positioned directly inside a sample tube, providing real-time control and monitoring of actual sample temperature.
MyBlock′s transparent lid helps to promote better uniformity and minimize temperature stratification in the block by maintaining a warm air jacket layer. The hinged lid can be flipped open (to the rear), or easily removed (lifted off) to accommodate taller tubes.
  • Digital temperature control (set and walk away)
  • Hinged lid (improves uniformity & conserves energy)
related product