Merck
1.17881
MAS-100 Sirius Flex Microbial Air Sampler
new
without perforated lid for all probable Flow Rates and Plate formats
Note: For More Information on this product, including pricing and availability, please click here!
Passer à
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
Matériaux
housing (stainless steel)
Niveau de qualité
Caractéristiques
portable
Fabricant/nom de marque
MAS-100® (Sirius)
dilution
(Air monitoring)
Application(s)
cosmetics
pharmaceutical
Description générale
The MAS-100 Sirius microbial air sampler is a portable, battery-operated device designed for active air sampling to collect and quantify microorganisms in ambient air. Utilizing an impaction principle similar to the Andersen air sampler, it simplifies environmental air monitoring with high efficiency, accuracy, and regulatory compliance.
The instrument boasts excellent recovery efficiency, defined by a nominal d50 value of 1.1 µm, with both physical and biological efficiencies validated according to ISO 14698 and EN 17141 for flow rates of 100 and 200 SLPM (standard Liters per minute) and accommodates various plate formats, including 55 mm contact plates and 90 mm settle plates.
The MAS-100 Sirius Flex is fully calibrated and offers flexibility without a perforated lid.
It features an easy plate exchange system with a magnetic closure and includes built-in, browser-based software compliant with 21 CFR Part 11, enabling seamless data transfer via barcode to LIMS or other
Application
The air sampler is designed for active air monitoring in cleanrooms acc. to current GMP guidance such as EU GMP Annex 1, FDA aseptic guide or PIC/S The instrument can be operated either innon-controlled areas as well as in cleanroomsenvironments up to grade A (or ISO 5) without compromising the viableand non-viable particle limits supported by an efficient exhaust HEPA filter.The housing is made of stainless steel with glass covered touchscreen forefficient cleaning and disinfection. The shape of the air path and air exhaustprevents turbulences of unidirectional air flow.
Caractéristiques et avantages
Excellent Performance
  • Independently validated biological & physical efficiency (acc. ISO 14698/EN 17141)
  • High accuracy and precision for selectable flow rates
Astonishing Safety features
  • Self checks
  • In-built leak test and dual mass flow sensor system
First Class cleanroom compliance
  • Easy to clean stainless steel surface
  • non-turbulent air flow path
  • HEPA exhaust filter
  • Active vaporized hydrogen peroxide decontamination
User friendly
  • Magnetic lid closure
  • Remote control and data transfer via Wi-Fi
Data integrity supporting features
  • Displays sampling barcode for seamless sample data transfer into LIMS
  • 21 CFR Part 11 compliant user management
  • Tamper proof audit trail
Autres remarques
Scope of Delivery:
1 x MAS-100 Sirius Microbial Air Sampler
1 x Blind Lid
1 x USB-C Cable (1.5 m)
1 x USB cover (mounted)
1 x Exhaust Filter (mounted)
1 x Transport case
Documents (Global Safety Data Sheet, clamp adjustment gauge, quick start guide, calibration certificates)
Please Note: the power supply (ref 1.17360) and perforated lid must be ordered separately.
Options for perforated Lids:
  • 1.17883: Suitable for 90 mm plates at a flow rate of 100 SLPM
  • 1.17884: Suitable for 55 mm plates at a flow rate of 100 SLPM
  • 1.17885: Suitable for 90 mm plates at a flow rate of 200 SLPM
  • 1.17886: Suitable for 55 mm plates at a flow rate of 200 SLPM
Informations légales
MAS-100 is a registered trademark of MBV AG
Articles
Discover the importance of data integrity in environmental monitoring. Learn how digital management systems can enhance compliance, safety, and efficiency in data handling.